3-Isocyanato-2-methyl-6-(trifluoromethyl)pyridine

Lieferant: Apollo Scientific
Achtung

PC300657-1G
APOSPC300657-1GEA 340 EUR
APOSPC300657-1G
3-Isocyanato-2-methyl-6-(trifluoromethyl)pyridine
3-Isocyanato-2-methyl-6-(trifluoromethyl)pyridine

When melted, may contain small amount of sediment

Formel: C₈H₅F₃N₂O
Molecular Weight: 202,133 g/mol
Siedepunkt: 144…146 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06808642
CAS-Nummer: 874832-10-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR