N-Benzyl-N'-[2-methyl-6-(trifluoromethyl)pyridin-3-yl]urea

Lieferant: Apollo Scientific

PC300660-1G
APOSPC300660-1GEA 0 EUR
APOSPC300660-1G
N-Benzyl-N'-[2-methyl-6-(trifluoromethyl)pyridin-3-yl]urea
N-Benzyl-N'-[2-methyl-6-(trifluoromethyl)pyridin-3-yl]urea
Formel: C₁₅H₁₄F₃N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD12406821
CAS-Nummer: 1227954-87-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR