4-Pyrrolidin-1-yl-2-(trifluoromethyl)aniline
Lieferant: Apollo Scientific
|
Achtung
|
PC300663-1G
APOSPC300663-1GEA
227
EUR
APOSPC300663-1G
4-Pyrrolidin-1-yl-2-(trifluoromethyl)aniline
4-Pyrrolidin-1-yl-2-(trifluoromethyl)aniline
Formel:
C₁₁H₁₃F₃N₂ Molecular Weight: 230.23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09050704 CAS-Nummer: 773899-18-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...