4-(4-Methyl-1-piperazinyl)-2-(trifluoromethyl)aniline
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(4-Methyl-1-piperazinyl)-2-(trifluoromethyl)aniline
PC300664-1G
PC300664-250MG
APOSPC300664-1GEA
190
EUR
APOSPC300664-1G
APOSPC300664-250MG
4-(4-Methyl-1-piperazinyl)-2-(trifluoromethyl)aniline
4-(4-Methyl-1-piperazinyl)-2-(trifluoromethyl)aniline
Formel:
C₁₂H₁₆F₃N₃ Molecular Weight: 259.28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09042650 CAS-Nummer: 863675-89-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...