N,2-Dimethyl-6-(trifluoromethyl)nicotinamide

Lieferant: Apollo Scientific

PC300669-1G
APOSPC300669-1GEA 232 EUR
APOSPC300669-1G
N,2-Dimethyl-6-(trifluoromethyl)nicotinamide
N,2-Dimethyl-6-(trifluoromethyl)nicotinamide
Formel: C₉H₉F₃N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD11826496
CAS-Nummer: 1090043-67-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR