2-Morpholin-4-yl-4-(trifluoromethyl)pyrimidine-5-carboxylic acid

Lieferant: Apollo Scientific

PC300673-1G
APOSPC300673-1GEA 359 EUR
APOSPC300673-1G
2-Morpholin-4-yl-4-(trifluoromethyl)pyrimidine-5-carboxylic acid
2-Morpholin-4-yl-4-(trifluoromethyl)pyrimidine-5-carboxylic acid
Formel: C₁₀H₁₀F₃N₃O₃
Molecular Weight: 277,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13188015
CAS-Nummer: 873450-23-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR