1-(4-Fluorophenoxy)-3-[4-(4-nitrophenyl)piperazin-1-yl]propan-2-ol

Lieferant: Apollo Scientific

PC300682-1G
APOSPC300682-1GEA 346 EUR
APOSPC300682-1G
1-(4-Fluorophenoxy)-3-[4-(4-nitrophenyl)piperazin-1-yl]propan-2-ol
1-(4-Fluorophenoxy)-3-[4-(4-nitrophenyl)piperazin-1-yl]propan-2-ol
Formel: C₁₉H₂₂FN₃O₄
Molecular Weight: 375,394 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15146813
CAS-Nummer: 1254122-84-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR