4-{[2-Methyl-6-(trifluoromethyl)pyridin-3-yl]amino}-4-oxobut-2-enoic acid

Lieferant: Apollo Scientific

PC300689-1G
APOSPC300689-1GEA 335 EUR
APOSPC300689-1G
4-{[2-Methyl-6-(trifluoromethyl)pyridin-3-yl]amino}-4-oxobut-2-enoic acid
4-{[2-Methyl-6-(trifluoromethyl)pyridin-3-yl]amino}-4-oxobut-2-enoic acid
Formel: C₁₁H₉F₃N₂O₃
Molecular Weight: 274,196 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01311984
CAS-Nummer: 402479-90-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR