3-[2-Methyl-6-(trifluoromethyl)pyridin-3-yl]acrylic acid

Lieferant: Apollo Scientific

PC300692-1G
APOSPC300692-1GEA 445 EUR
APOSPC300692-1G
3-[2-Methyl-6-(trifluoromethyl)pyridin-3-yl]acrylic acid
3-[2-Methyl-6-(trifluoromethyl)pyridin-3-yl]acrylic acid
Formel: C₁₀H₈F₃NO₂
Molecular Weight: 231.171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20037417
CAS-Nummer: 545394-86-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR