4-Bromo-3-fluoro-N-(tetrahydrofuran-2-ylmethyl)benzenesulphonamide

Lieferant: Apollo Scientific

PC300694-1G
APOSPC300694-1GEA 296 EUR
APOSPC300694-1G
4-Bromo-3-fluoro-N-(tetrahydrofuran-2-ylmethyl)benzenesulphonamide
4-Bromo-3-fluoro-N-(tetrahydrofuran-2-ylmethyl)benzenesulphonamide
Formel: C₁₁H₁₃BrFNO₃S
Molecular Weight: 338,193 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18804166
CAS-Nummer: 1207256-50-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR