{5-[3,5-Di(trifluoromethyl)phenyl]-2-furyl}methanol

Lieferant: Apollo Scientific

PC300695-1G
APOSPC300695-1GEA 125 EUR
APOSPC300695-1G
{5-[3,5-Di(trifluoromethyl)phenyl]-2-furyl}methanol
{5-[3,5-Di(trifluoromethyl)phenyl]-2-furyl}methanol
Formel: C₁₃H₈F₆O₂
Molecular Weight: 310,192 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00052573
CAS-Nummer: 424803-19-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR