2-Methyl-5-[2-(trifluoromethoxy)phenyl]-1,3,4-oxadiazole

Lieferant: Apollo Scientific

PC300698-1G
APOSPC300698-1GEA 206 EUR
APOSPC300698-1G
2-Methyl-5-[2-(trifluoromethoxy)phenyl]-1,3,4-oxadiazole
2-Methyl-5-[2-(trifluoromethoxy)phenyl]-1,3,4-oxadiazole
Formel: C₁₀H₇F₃N₂O₂
Molecular Weight: 244,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20441492
CAS-Nummer: 1427460-89-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR