6-(Trifluoromethyl)-1-benzothiophene-2-carbohydrazide

Lieferant: Apollo Scientific
Achtung

PC300701-1G
APOSPC300701-1GEA 21.9 EUR
APOSPC300701-1G
6-(Trifluoromethyl)-1-benzothiophene-2-carbohydrazide
6-(Trifluoromethyl)-1-benzothiophene-2-carbohydrazide
Formel: C₁₀H₇F₃N₂OS
Molecular Weight: 260.24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11615820
CAS-Nummer: 1135282-94-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR