4-(Trifluoromethyl)benzo[b]thiophene-2-carbohydrazide

Lieferant: Apollo Scientific
Achtung

PC300702-1G
APOSPC300702-1GEA 395 EUR
APOSPC300702-1G
4-(Trifluoromethyl)benzo[b]thiophene-2-carbohydrazide
4-(Trifluoromethyl)benzo[b]thiophene-2-carbohydrazide
Formel: C₁₀H₇F₃N₂OS
Lagertemperatur: Raumtemperatur
MDL: MFCD11615821
CAS-Nummer: 1171927-41-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR