3-Bromo-7-(trifluoromethyl)-1H-indazole

Lieferant: Apollo Scientific
Achtung

PC300707-1G
APOSPC300707-1GEA 433 EUR
APOSPC300707-1G
3-Bromo-7-(trifluoromethyl)-1H-indazole
3-Bromo-7-(trifluoromethyl)-1H-indazole
Formel: C₈H₄BrF₃N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11848410
CAS-Nummer: 885693-99-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR