3-(Trifluoromethoxy)benzamidoxime
Lieferant: Apollo Scientific
|
Achtung
|
PC300708-1G
PC300708-5G
PC300708-25G
APOSPC300708-1GEA
0
EUR
APOSPC300708-1G
APOSPC300708-5G
APOSPC300708-25G
3-(Trifluoromethoxy)benzamidoxime
3-(Trifluoromethoxy)benzamidoxime
Formel:
C₈H₇F₃N₂O₂ Schmelzpunkt: 50…52 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06660166 CAS-Nummer: 886500-80-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...