5-[4-(Trifluoromethoxy)phenyl]-1,3-oxazole

Lieferant: Apollo Scientific

PC300709-1G
APOSPC300709-1GEA 213 EUR
APOSPC300709-1G
5-[4-(Trifluoromethoxy)phenyl]-1,3-oxazole
5-[4-(Trifluoromethoxy)phenyl]-1,3-oxazole
Formel: C₁₀H₆F₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD12406837
CAS-Nummer: 1227954-44-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR