1-(2-fluoro-4-nitrophenyl)piperidin-4-one

Lieferant: Apollo Scientific

PC300710-1G
APOSPC300710-1GEA 241 EUR
APOSPC300710-1G
1-(2-fluoro-4-nitrophenyl)piperidin-4-one
1-(2-fluoro-4-nitrophenyl)piperidin-4-one
Formel: C₁₁H₁₁FN₂O₃
Lagertemperatur: Raumtemperatur
CAS-Nummer: 439097-58-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR