2-Amino-4-(trifluoromethyl)benzhydrazide
Lieferant: Apollo Scientific
PC300716-1G
APOSPC300716-1GEA
241
EUR
APOSPC300716-1G
2-Amino-4-(trifluoromethyl)benzhydrazide
2-Amino-4-(trifluoromethyl)benzhydrazide
Formel:
C₈H₈F₃N₃O Lagertemperatur: Raumtemperatur |
MDL:
MFCD18804173 CAS-Nummer: 1228794-36-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...