5-Chloro-2-fluorobenzhydrazide

Lieferant: Apollo Scientific
Achtung

PC300719-1G
APOSPC300719-1GEA 306 EUR
APOSPC300719-1G
5-Chloro-2-fluorobenzhydrazide
5-Chloro-2-fluorobenzhydrazide
Formel: C₇H₆ClFN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD09816933
CAS-Nummer: 657424-44-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR