2-(Chloromethyl)-5-(2-fluorophenyl)-1,3,4-oxadiazole
Lieferant: Apollo Scientific
|
Achtung
|
PC300720-1G
APOSPC300720-1GEA
319
EUR
APOSPC300720-1G
2-(Chloromethyl)-5-(2-fluorophenyl)-1,3,4-oxadiazole
2-(Chloromethyl)-5-(2-fluorophenyl)-1,3,4-oxadiazole
Formel:
C₉H₆ClFN₂O Molecular Weight: 212,608 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09041034 CAS-Nummer: 350672-17-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...