N-(4-Methoxyphenyl)-N'-[2-methyl-6-(trifluoromethyl)pyridin-3-yl]urea

Lieferant: Apollo Scientific

PC300728-1G
APOSPC300728-1GEA 601 EUR
APOSPC300728-1G
N-(4-Methoxyphenyl)-N'-[2-methyl-6-(trifluoromethyl)pyridin-3-yl]urea
N-(4-Methoxyphenyl)-N'-[2-methyl-6-(trifluoromethyl)pyridin-3-yl]urea
Formel: C₁₅H₁₄F₃N₃O₂
Molecular Weight: 325,286 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12406828
CAS-Nummer: 1227954-51-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR