N-(2-Methoxyphenyl)-N'-[2-methyl-6-(trifluoromethyl)pyridin-3-yl]urea
Lieferant: Apollo Scientific
PC300732-1G
APOSPC300732-1GEA
601
EUR
APOSPC300732-1G
N-(2-Methoxyphenyl)-N'-[2-methyl-6-(trifluoromethyl)pyridin-3-yl]urea
N-(2-Methoxyphenyl)-N'-[2-methyl-6-(trifluoromethyl)pyridin-3-yl]urea
Formel:
C₁₅H₁₄F₃N₃O₂ Molecular Weight: 325,286 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12406832 CAS-Nummer: 1227954-80-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...