2-Methyl-6-(trifluormethyl)nicotinsäurehydrazid
Lieferant: Apollo Scientific
PC300733-1G
APOSPC300733-1GEA
109
EUR
APOSPC300733-1G
2-Methyl-6-(trifluormethyl)nicotinsäurehydrazid
2-Methyl-6-(trifluormethyl)nicotinsäurehydrazid
Formel:
C₈H₈F₃N₃O Lagertemperatur: Raumtemperatur |
MDL:
MFCD01565843 CAS-Nummer: 402479-94-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...