4-(4-Morpholinyl)-2-(trifluoromethyl)aniline

Lieferant: Apollo Scientific
Achtung

PC300734-250MG
APOSPC300734-250MGEA 0 EUR
APOSPC300734-250MG
4-(4-Morpholinyl)-2-(trifluoromethyl)aniline
4-(4-Morpholinyl)-2-(trifluoromethyl)aniline
Formel: C₁₁H₁₃F₃N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD01436021
CAS-Nummer: 78335-26-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR