3-Bromo-8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

PC300738-250MG
APOSPC300738-250MGEA 383 EUR
APOSPC300738-250MG
3-Bromo-8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid
3-Bromo-8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid
Formel: C₉H₄BrF₃N₂O₂
Molecular Weight: 309,039 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17168281
CAS-Nummer: 1355171-41-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR