3-Chloro-8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC300739-100MG
APOSPC300739-100MGEA
325
EUR
APOSPC300739-100MG
3-Chloro-8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid
3-Chloro-8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid
Formel:
C₉H₄ClF₃N₂O₂ Molecular Weight: 264,588 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD17168283 CAS-Nummer: 1237838-81-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...