3-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

PC300740-1G PC300740-250MG
APOSPC300740-1GEA 34.8 EUR
APOSPC300740-1G APOSPC300740-250MG
3-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid
3-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid
Formel: C₉H₄ClF₃N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD17168284
CAS-Nummer: 1355171-48-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR