6-Bromo-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC300742-100MG
APOSPC300742-100MGEA
85.7
EUR
APOSPC300742-100MG
6-Bromo-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid
6-Bromo-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid
Formel:
C₉H₄BrF₃N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD18804153 CAS-Nummer: 1427460-57-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...