2-[(Dimethylamino)methylene]-6-fluoroindan-1-one

Lieferant: Apollo Scientific
Achtung

PC300746-100MG
APOSPC300746-100MGEA 21.9 EUR
APOSPC300746-100MG
2-[(Dimethylamino)methylene]-6-fluoroindan-1-one
2-[(Dimethylamino)methylene]-6-fluoroindan-1-one
Formel: C₁₂H₁₂FNO
Molecular Weight: 205,228 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19442756
CAS-Nummer: 1427462-76-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR