N-Methyl-4-(trifluoromethoxy)benzenesulphonamide

Lieferant: Apollo Scientific
Achtung

PC300748-100MG
APOSPC300748-100MGEA 21.9 EUR
APOSPC300748-100MG
N-Methyl-4-(trifluoromethoxy)benzenesulphonamide
N-Methyl-4-(trifluoromethoxy)benzenesulphonamide
Formel: C₈H₈F₃NO₃S
Molecular Weight: 255.214 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08599121
CAS-Nummer: 909527-60-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR