8-Bromo-6-fluoroimidazo[1,2-a]pyridine
Lieferant: Apollo Scientific
|
Achtung
|
PC300754-1G
PC300754-250MG
APOSPC300754-1GEA
166
EUR
APOSPC300754-1G
APOSPC300754-250MG
8-Bromo-6-fluoroimidazo[1,2-a]pyridine
8-Bromo-6-fluoroimidazo[1,2-a]pyridine
Formel:
C₇H₄BrFN₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD20923664 CAS-Nummer: 1368664-08-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...