8-Bromo-6-fluoroimidazo[1,2-a]pyridine

Lieferant: Apollo Scientific
Achtung

PC300754-1G PC300754-250MG
APOSPC300754-1GEA 166 EUR
APOSPC300754-1G APOSPC300754-250MG
8-Bromo-6-fluoroimidazo[1,2-a]pyridine
8-Bromo-6-fluoroimidazo[1,2-a]pyridine
Formel: C₇H₄BrFN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD20923664
CAS-Nummer: 1368664-08-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR