2-{4-[5-(Trifluoromethyl)-1,3,4-thiadiazol-2-yl]piperazin-1-yl}ethanol

Lieferant: Apollo Scientific

PC300755-1G
APOSPC300755-1GEA 296 EUR
APOSPC300755-1G
2-{4-[5-(Trifluoromethyl)-1,3,4-thiadiazol-2-yl]piperazin-1-yl}ethanol
2-{4-[5-(Trifluoromethyl)-1,3,4-thiadiazol-2-yl]piperazin-1-yl}ethanol
Formel: C₉H₁₃F₃N₄OS
Molecular Weight: 282,286 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25976391
CAS-Nummer: 1713589-54-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR