2,3,5-Trifluoro-4-hydrazinobenzhydrazide

Lieferant: Apollo Scientific

Synonyme: 2,3,5-Trifluoro-4-hydrazinobenzohydrazide

PC300757-1G
APOSPC300757-1GEA 361 EUR
APOSPC300757-1G
2,3,5-Trifluoro-4-hydrazinobenzhydrazide
2,3,5-Trifluoro-4-hydrazinobenzhydrazide
Formel: C₇H₇F₃N₄O
Molecular Weight: 220,152 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27995621
CAS-Nummer: 1823962-17-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR