Ethyl 2,3,5-trifluoro-4-hydroxybenzoate

Lieferant: Apollo Scientific
Achtung

PC300759-1G
APOSPC300759-1GEA 319 EUR
APOSPC300759-1G
Ethyl 2,3,5-trifluoro-4-hydroxybenzoate
Ethyl 2,3,5-trifluoro-4-hydroxybenzoate
Formel: C₉H₇F₃O₃
Molecular Weight: 220,145 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14698251
CAS-Nummer: 1214351-42-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR