Methyl 2,3,5-trifluoro-4-methoxybenzoate

Lieferant: Apollo Scientific
Achtung

PC300760-1G PC300760-5G
APOSPC300760-1GEA 364 EUR
APOSPC300760-1G APOSPC300760-5G
Methyl 2,3,5-trifluoro-4-methoxybenzoate
Methyl 2,3,5-trifluoro-4-methoxybenzoate
Formel: C₉H₇F₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD16877111
CAS-Nummer: 1656263-44-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR