Methyl 2,3,5-trifluoro-4-morpholin-4-ylbenzoate

Lieferant: Apollo Scientific

PC300761-1G PC300761-250MG
APOSPC300761-1GEA 232 EUR
APOSPC300761-1G APOSPC300761-250MG
Methyl 2,3,5-trifluoro-4-morpholin-4-ylbenzoate
Methyl 2,3,5-trifluoro-4-morpholin-4-ylbenzoate
Formel: C₁₂H₁₂F₃NO₃
Molecular Weight: 275.224 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27995633
CAS-Nummer: 1858250-71-1

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR