Methyl 2,3,5-trifluoro-4-piperidin-1-ylbenzoate

Lieferant: Apollo Scientific

PC300762-1G
APOSPC300762-1GEA 232 EUR
APOSPC300762-1G
Methyl 2,3,5-trifluoro-4-piperidin-1-ylbenzoate
Methyl 2,3,5-trifluoro-4-piperidin-1-ylbenzoate
Formel: C₁₃H₁₄F₃NO₂
Molecular Weight: 273,251 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28040510
CAS-Nummer: 1858249-59-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR