5-(4-Fluoro-3-iodophenyl)-1,3-oxazole

Lieferant: Apollo Scientific
Achtung

PC300765-1G
APOSPC300765-1GEA 478 EUR
APOSPC300765-1G
5-(4-Fluoro-3-iodophenyl)-1,3-oxazole
5-(4-Fluoro-3-iodophenyl)-1,3-oxazole
Formel: C₉H₅FINO
Molecular Weight: 289,045 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27995623
CAS-Nummer: 1823893-90-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR