N-[4-Bromo-2-(trifluoromethoxy)phenyl]-N'-(1-tetrahydro-2H-pyran-4-yl-1H-pyrazol-4-yl)thiourea

Lieferant: Apollo Scientific

PC300767-1G
APOSPC300767-1GEA 371 EUR
APOSPC300767-1G
N-[4-Bromo-2-(trifluoromethoxy)phenyl]-N'-(1-tetrahydro-2H-pyran-4-yl-1H-pyrazol-4-yl)thiourea
N-[4-Bromo-2-(trifluoromethoxy)phenyl]-N'-(1-tetrahydro-2H-pyran-4-yl-1H-pyrazol-4-yl)thiourea
Formel: C₁₆H₁₆BrF₃N₄O₂S
Molecular Weight: 465,288 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27995616
CAS-Nummer: 1858250-66-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR