3-Iodo-2-methyl-6-(trifluoromethyl)pyridine

Lieferant: Apollo Scientific
Achtung

PC300768-1G
APOSPC300768-1GEA 546 EUR
APOSPC300768-1G
3-Iodo-2-methyl-6-(trifluoromethyl)pyridine
3-Iodo-2-methyl-6-(trifluoromethyl)pyridine
Formel: C₇H₅F₃IN
Molecular Weight: 287,021 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27934112
CAS-Nummer: 944317-26-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR