2-[2-(6-Fluoroimidazo[1,2-a]pyridin-2-yl)ethyl]-1H-isoindole-1,3(2H)-dione hydrobromide

Lieferant: Apollo Scientific

PC300769-1G
APOSPC300769-1GEA 371 EUR
APOSPC300769-1G
2-[2-(6-Fluoroimidazo[1,2-a]pyridin-2-yl)ethyl]-1H-isoindole-1,3(2H)-dione hydrobromide
2-[2-(6-Fluoroimidazo[1,2-a]pyridin-2-yl)ethyl]-1H-isoindole-1,3(2H)-dione hydrobromide
Formel: C₁₇H₁₂FN₃O₂·BrH
Lagertemperatur: Raumtemperatur
MDL: MFCD27995622
CAS-Nummer: 1858250-77-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR