5-(6-Fluoro-4H-1,3-benzodioxin-8-yl)-1,3-oxazole
Lieferant: Apollo Scientific
PC300771-1G
APOSPC300771-1GEA
371
EUR
APOSPC300771-1G
5-(6-Fluoro-4H-1,3-benzodioxin-8-yl)-1,3-oxazole
5-(6-Fluoro-4H-1,3-benzodioxin-8-yl)-1,3-oxazole
Formel:
C₁₁H₈FNO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD27995625 CAS-Nummer: 1823272-05-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...