4-[2-Methyl-6-(trifluoromethyl)pyridin-3-yl]pyrimidine-2-thiol

Lieferant: Apollo Scientific

PC300772-1G
APOSPC300772-1GEA 420 EUR
APOSPC300772-1G
4-[2-Methyl-6-(trifluoromethyl)pyridin-3-yl]pyrimidine-2-thiol
4-[2-Methyl-6-(trifluoromethyl)pyridin-3-yl]pyrimidine-2-thiol
Formel: C₁₁H₈F₃N₃S
Molecular Weight: 271,262 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27995628
CAS-Nummer: 1858250-86-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR