3-(Bromoacetyl)-2-methyl-6-(trifluoromethyl)pyridine hydrobromide
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromo-1-[2-methyl-6-(trifluoromethyl)pyridin-3-yl]ethan-1-one hydrobromide
PC300773-1G
PC300773-250MG
PC300773-5G
APOSPC300773-1GEA
226
EUR
APOSPC300773-1G
APOSPC300773-250MG
APOSPC300773-5G
3-(Bromoacetyl)-2-methyl-6-(trifluoromethyl)pyridine hydrobromide
3-(Bromoacetyl)-2-methyl-6-(trifluoromethyl)pyridine hydrobromide
Formel:
C₉H₇BrF₃NO·BrH Molecular Weight: 362.969 g/mol Schmelzpunkt: 145…150 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD28040497 CAS-Nummer: 1377962-47-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...