4-[2-Methyl-6-(trifluoromethyl)pyridin-3-yl]-1,3-thiazol-2-amine

Lieferant: Apollo Scientific

PC300774-1G
APOSPC300774-1GEA 371 EUR
APOSPC300774-1G
4-[2-Methyl-6-(trifluoromethyl)pyridin-3-yl]-1,3-thiazol-2-amine
4-[2-Methyl-6-(trifluoromethyl)pyridin-3-yl]-1,3-thiazol-2-amine
Formel: C₁₀H₈F₃N₃S
Molecular Weight: 259,251 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27995632
CAS-Nummer: 1858241-52-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR