2-Phenyl-6-trifluoromethyl-nicotinic acid ethyl ester

Lieferant: Apollo Scientific
Achtung

PC300775-1G
APOSPC300775-1GEA 371 EUR
APOSPC300775-1G
2-Phenyl-6-trifluoromethyl-nicotinic acid ethyl ester
2-Phenyl-6-trifluoromethyl-nicotinic acid ethyl ester
Formel: C₁₅H₁₂F₃NO₂
Molecular Weight: 295,256 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11113427
CAS-Nummer: 149770-27-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR