6-(Trifluoromethyl)imidazo[1,2-a]pyridin-3-amine

Lieferant: Apollo Scientific
Achtung

PC300776-1G
APOSPC300776-1GEA 1810 EUR
APOSPC300776-1G
6-(Trifluoromethyl)imidazo[1,2-a]pyridin-3-amine
6-(Trifluoromethyl)imidazo[1,2-a]pyridin-3-amine
Formel: C₈H₆F₃N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD22076853
CAS-Nummer: 1536950-06-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR