2-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}benzoic acid

Lieferant: Apollo Scientific

PC300779-1G PC300779-5G
APOSPC300779-1GEA 0 EUR
APOSPC300779-1G APOSPC300779-5G
2-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}benzoic acid
2-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}benzoic acid
Formel: C₁₃H₇ClF₃NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD03644663
CAS-Nummer: 852952-20-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR